Floor Pad, Eco-Prep, designed to be used wet or dry for the removal of floor finish without the use of harsh stripping chemicals, Excellent for repairing acrylic or urethane finishes for recoating, Machine speed: up to 350 RPM, Maroon, 20in
# de catalogue 76478-704
Fournisseur: Medline
Customers Who Bought This Also Bought