Media/Storage Bottles, United Scientific
# de catalogue 470332-672
Fournisseur: United Scientific Supplies
Spécifications
- Capacity:2000 ml
- Description:Media/storage bottles
- Cat. no.:470332-672
Spécifications
A propos de cet article
General-purpose media bottles.
- Autoclavable
- Borosilicate glass bottles
- Supplied with polypropylene cap
These borosilicate glass general-purpose media bottles are autoclavable with permanent white imprinted graduations and marking spots. The screw thread opening has an int.Ø of approximately 30 mm. The supplied GL 45 cap is linerless, and both the cap and drip-free ring are polypropylene and autoclavable to 140 °C. Maximum use temperature of the bottles is 450 °C.
Customers Who Bought This Also Bought