ContactUsLinkComponent
Rubber feet reduce the movement of Foundation platforms that are 12 feet (3.66 m) and under. Simply slide the leg cross brace up and place a foot on the bottom of each leg, No tools required, Black 2in Thickness, 2in Dimension
# de catalogue 76475-004
Fournisseur:  Wearwell
Rubber feet reduce the movement of Foundation platforms that are 12 feet (3.66 m) and under. Simply slide the leg cross brace up and place a foot on the bottom of each leg, No tools required, Black 2in Thickness, 2in Dimension
# de catalogue 76475-004
Fournisseur:  Wearwell
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.