Kit, Esd-Audit, Metriso B530, with Walking Test, include instrument/equipment needed to perform ESD auditing according to IEC 61340-5-1/ qualification/verification of ESD control element, Evaluating temperature/relative humidity, Evaluating Ionizer acc to IEC61340-4-7, Outer dimension: 575x490x250mm
# de catalogue 76474-642
Fournisseur: Transforming Technologies
Customers Who Bought This Also Bought