Kit, Esd-Audit Metriso B530, includes instruments and equipment needed to perform ESD auditing according to IEC 61340-4-1 and qualification/ verification of ESD control elements, Evaluating temperature and relative humidity, Evaluating Ionizers acc. To IEC 61340-4-7, Outer dimension: 575x490 x250mm
# de catalogue 76474-640
Fournisseur: Transforming Technologies
Customers Who Bought This Also Bought