ZymoBIOMICS Fecal Reference with TruMatrix Technology, microbial reference material composed of stool from healthy donors for quality control, process validation, assay development, and proficiency testing, 1mL of the product contains about 10 mg feces, 10 preps
# de catalogue 76474-118
Fournisseur: Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.