Spécifications
- Length:44.6 mm
- Volume:10 µl
- Tip Color:Clear
- Packed:10 Racks, 96 Tips each
- Filter:+
- Sterile:+
- Tip Style:Universal, low retention
Spécifications
A propos de cet article
Azer Scientific’s ExacTip™ pipette tip line is the ideal choice for a universal pipette tip or universal filtered pipette tip. The surface and material of the filter tips utilize special processes and polypropylene resins to ensure the low retention qualities for ensuring they are one of the most accurate pipette tips in the market.
- Manufactured in a Class 100,000 cleanroom and sterilized under ISO 11137 guidelines
- Tips have low DNA and protein binding treatment
- DNase, RNase, and Pyrogren free
- Sterile individually wrapped racks of 96 tips
- Contain small micron hydrophobic barrier filters to prevent contaminating aerosols from entering your pipette
These filtered pipette tips are manufactured under stringent ISO 13485:2016 and ISO 9001:2015 quality control guidelines in state-of-the-art Class 100,000 cleanroom environments.
Universal filtered pipette tips give you the flexibility of pipette choice, eliminating the need to purchase expensive brand-specific tips for your pipettor. The manufacturing and sterilization processes allow for the pipette tips to be used for many laboratory applications including molecular biology, cell culture, PCR, toxicology and much more.