Digital Dispenser, Pico 1, Voltage: 100 to 240V 50/60Hz, For Use With Drug combination studies, Biochemical assays, Cell-based assays, Interface: USB, Dispensing Range: 11 pL to 20 uL or 1 nL to 200 uL, Dimensions (L x W x H): 31 cm x 28 cm x 20 cm
# de catalogue 76473-008
Fournisseur: Thermo Fisher Scientific
Customers Who Bought This Also Bought