ContactUsLinkComponent
Vactrap2™ Vacuum Trap System Vacuum Protection, with Secondary Container and Inline Vent Filter, Foxx Life Sciences
Catalogue # 76472-596
Fournisseur:  Foxx Life Sciences
undefined
Vactrap2™ Vacuum Trap System Vacuum Protection, with Secondary Container and Inline Vent Filter, Foxx Life Sciences
Catalogue # 76472-596
Fournisseur:  Foxx Life Sciences
Supplier Number:  302-1101-FLS
Spécifications
  • Description:
    Vacuum trap system, shatterproof aspirator bottle assembly with secondary container and inline hydrophobic vent filter, with overflow bottle, HDPE, 1 L + 1 L overflow bottle
  • Cat. no.:
    76472-596
Spécifications
À propos de cet article
Vactrap2™ Vacuum protection system (vacuum flask alternative) for aspiration and vacuum pump protection.
  • Vactrap2™ is available in autoclavable PP or chemically-resistant HDPE
  • Each Vactrap2™ includes the unique VersaCap® closure for tangle-free opening and closing
  • 0.2 μm PTFE inline vent filter retains any aerosolized bacteria and protects the house vacuum or pump
  • The entire assembly is nested neatly in a secondary container with a small footprint and incredible stability against tipping
  • Pinch clamp allows for vacuum adjustment
Vactrap™ 2 is an advanced vacuum trap system. Ideal for the protection of vacuum lines and scientists alike, Vactrap2™ safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap2™ comes ready to use; simply attach to your vacuum and begin your work.
The full Vactrap2™  system includes two Foxx Heavy Duty Vacuum Bottles (autoclavable PP or chemically-resistant HDPE), each with a unique VersaCap® closure for tangle-free opening, no need to disconnect tubing, ever. New, improved heavy-duty EZBio® TPE V tubing to avoid kinking. A 0.2 µm PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nestled neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for the modulation of vacuum. Other Vactrap™ 2 options offer one bottle or setups without secondary containment. 
Certifications: 40 CFR 264.175 compliant.
Frequently Bought Together