ContactUsLinkComponent
Pipette Controller, Variable aliquoting speed - customer adjustable with 7 speeds, Aliquoting speed control dial, On-board calibration, Removable kickstand, Repeat pipetting plus conventional pipetting with a single 2-in-1 controller, Use any size and brand of serological pipets, Size: 0.5 to 5 mL
# de catalogue MP76382-930
Fournisseur:  VistaLab
Pipette Controller, Variable aliquoting speed - customer adjustable with 7 speeds, Aliquoting speed control dial, On-board calibration, Removable kickstand, Repeat pipetting plus conventional pipetting with a single 2-in-1 controller, Use any size and brand of serological pipets, Size: 0.5 to 5 mL
# de catalogue MP76382-930
Fournisseur:  VistaLab
Customers Who Bought This Also Bought