Cartridge, spe, Resprep Diol, (Mixed-Mode phase), High-quality, silica-based adsorbents, Used for common sample matrices such as food and water, Stable over the pH range (2 - 8), Size: 6mL/1000mg
# de catalogue 76470-258
Fournisseur: Restek
Customers Who Bought This Also Bought