Cartridge, spe, Resprep SAX (Silica Based), (Ion-Exchange mode), High-quality, silica-based hydrophilic adsorbents, Used for common sample matrices such as food, soil, and water, SAX - strong anion exchange, Stable over the pH range (2 - 9), Size: 6mL/500mg
# de catalogue 76470-250
Fournisseur: Restek
Customers Who Bought This Also Bought