Cartridge, spe, Resprep PSA, Highquality, silica-based adsorbents, Used for common sample matrices such as food, soil, water, and petroleum, PSA - primary secondary amine, In addition to primary use mechanisms(reversed-phase or normal phase), PSA may be used as a weak anion exchange, Size: 6mL/500mg
# de catalogue 76470-248
Fournisseur: Restek
Customers Who Bought This Also Bought