Cartridge, spe, Resprep Amino (NH2), High-quality, silica-based adsorbents, Used for common sample matrices such as food, soil, and petroleum, In addition to its primary use mechanisms (reversed-phase or normal phase), NH2 may also be used as a weak anion exchange (WAX), Size: 6mL/500mg
# de catalogue 76470-244
Fournisseur: Restek
Customers Who Bought This Also Bought