Connector, Sanitary Genderless Aseptiquik G, Material: Polycarbonate, USP Class VI, ADCF, Color: White with blue pull tab, Material Finish: Molded White, Free Floating, Gamma sterilization, Temperature Range: 34 deg F - 104 deg F (1 deg C - 40 deg C), Non-Valved, Volume: 3/4 In
# de catalogue 76470-190
Fournisseur: Colder Products
Customers Who Bought This Also Bought