ContactUsLinkComponent
Filter Pipette Tip, Plain, Certified, Universal, Low Retention, Graduated, Natural, 84mm, Extended Length, sterile, Size: 1 - 1000uL
Sterile Low Retention Filter Pipette Tips, Globe Scientific
Catalog # 76468-288
Supplier:  Globe Scientific
Filter Pipette Tip, Plain, Certified, Universal, Low Retention, Graduated, Natural, 84mm, Extended Length, sterile, Size: 1 - 1000uL
Sterile Low Retention Filter Pipette Tips, Globe Scientific
Catalog # 76468-288
Supplier:  Globe Scientific
Specifications
  • Volume:
    1 - 1000 µl
  • Length:
    86 mm
  • Packed:
    96 per Rack, 20 Racks per case
  • Cat. no.:
    76468-288
  • Sterile:
    +
  • Tip Style:
    Extended length
Specifications
About this item
Universal fit is compatible with most common brands of pipettors.
  • Manufactured from clear polypropylene (PP)
  • Polypropylene (PP) filters protect samples and pipettors from contamination
  • DNase-free, RNase-free and non-pyrogenic
  • Manufactured in a Class 100 000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems
  • Sterilized by gamma irradiation SAL 10⁻⁶ (ISO 11137)
  • Racks are printed with lot number for quality traceability
Special processing renders these tips hydrophobic for exceptionally low sample retention. A universal fit design provides an airtight fit on most brands of pipettors.
Customers Who Bought This Also Bought