Sterile Low Retention Filter Pipette Tips, Globe Scientific
# de catalogue 76468-284
Fournisseur: Globe Scientific
Spécifications
- Length:59 mm
- Volume:1 - 200 µl
- Packed:96 per Rack, 20 Racks per case
- Sterile:+
- Tip Style:Standard
Spécifications
A propos de cet article
Universal fit is compatible with most common brands of pipettors.
- Manufactured from clear polypropylene (PP)
- Polypropylene (PP) filters protect samples and pipettors from contamination
- DNase-free, RNase-free and non-pyrogenic
- Manufactured in a Class 100 000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems
- Sterilized by gamma irradiation SAL 10⁻⁶ (ISO 11137)
- Racks are printed with lot number for quality traceability
Special processing renders these tips hydrophobic for exceptionally low sample retention. A universal fit design provides an airtight fit on most brands of pipettors.
Customers Who Bought This Also Bought