Pipet tip, Robotic filter, for automation, Hamilton compatible, conductive 50ul filter tips, Color: Black, Tip Shape: Round, Polypropylene, Nonpyrogenic, Non-sterile, Liquid Level Sensing Tips, 96/rack
# de catalogue 76463-400
Fournisseur: Azer Scientific Inc.
Customers Who Bought This Also Bought