Face mask, Earloop, Level 2, Single pleats rectangular face masks with a shapeable nosepiece and two earloops present, one on each side, in order to hold mask in place, Outer Material Spunbond Polypropylene (SBPP), Regular length, Non sterile, Length 175 mm (6-7/8in)
# de catalogue 76461-930
Fournisseur: Medicom
Customers Who Bought This Also Bought