AssureWear™ SMS Isolation Gowns, Medicom
# de catalogue 76461-928
Fournisseur: Medicom
Spécifications
- Clothing Size:L
- Garment application:Medical
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Back fastening
- Color:Yellow
- Cat. no.:76461-928
Spécifications
A propos de cet article
Provides reliable protection without compromising comfort.
- Single-use gown
- AAMI Level 2
- Features neck ties, elastic cuffs, and full back closure
- Sewn seams ensure superior strength
- Latex-free
These 3-layer SMS Isolation gowns feature a tape tab neck, elastic cuffs, and full back. Single-use gowns are designed to provide reliable protection without compromising comfort. Sewn seams ensure superior strength.
Disposable SMMS Polypropylene Isolation Gown designed to protect health professionals and patients from cross infection.
Packaging: Gowns are packaged 10/bag, 10 bags/case.
Customers Who Bought This Also Bought