VWR® EZLabpure™ Round Carboys, with 83B VersaCap®
Catalog # 76461-402
Supplier: VWR International
Specifications
- Cap Type:VersaCap® 83B
- Capacity:10 L
- Description:EZLabpure™ round carboy, with spigot
- Material:PP
- Cat. no.:76461-402
- Diameter ×H:248×392 mm
Specifications
About this item
VWR® EZLabpure™ round carboys are manufactured using high-purity virgin USP Class VI medical grade polypropylene.
- Carboys can be used for storing and shipping reagents, chemicals, culture media, and standards
- Polypropylene carboys are autoclavable at 121 °C for twenty minutes
- Can be used at a temperature below 0 °C
- Superior chemical resistance
A premium line of high-quality, round HDPE and polypropylene (PP) laboratory carboys with the patented VersaCap® technology. These durable, rugged, and shatterproof round carboys are a great alternative to glass carboys. They can be used for mixing, storing, and dispensing liquids in volumes from 10 to 50 liters. These carboys are available with and without a spigot.
Certifications: UPS Class VI materials.
Customers Who Bought This Also Bought