Spécifications
- Ø:12/6 mm
- Description:Single junction, ceramic diaphram
- Material:Glass
- Cat. no.:76460-480
- Connector:BNC, 1 m cable
- Type:SM 223 (semi-micro)
- Electrolyte:KCl 3 mol/L
Spécifications
A propos de cet article
Laboratory and field electrodes for general applications. Supplied with a fixed cable or plug head, VWR® electrodes can be connected to most pH meters on the market.
- Comfortable handling
- Reliable, rapid readings
- Gel or liquid electrolyte, glass or epoxy body shaft
- Available with different connections: Plug head or fixed cable (1 metre) with either DIN or BNC type plug
All VWR electrodes are checked at two points during the manufacturing process and then lot stamped to ensure quality. They are supplied with a soaker storage bottle and cap to make sure the pH-sensitive glass and reference junction remain in top condition, and are instantly ready for use.