Order Entry
ContactUsLinkComponent
Coaxial Insert, for 5 mm NMR Precision Sample Tube, Larger Inner Volume, most versatile and reliable coaxial system available for various NMR experiments, Ideal for variable temperature studies, all parts are made from glass, Dimension: 50x3.3mm, Volume: 220x260ul, Use with: 506-PP to 535-PP
SP Wilmad-LabGlass Stem Coaxial Small Volume NMR Inserts, SP Industries
# de catalogue 76460-278
Fournisseur:  SP Industries
SP Wilmad-LabGlass Stem Coaxial Small Volume NMR Inserts, SP Industries
# de catalogue 76460-278
Fournisseur:  SP Industries

Spécifications

  • Length:
    50 mm (stem)
  • O.D.:
    3.3 mm (stem)
  • Volume:
    220/260 µl (inner/outer)
  • For Use With:
    Tubes with Ext.Ø. 5 mm

Spécifications

A propos de cet article

The most versatile and reliable coaxial system available for NMR experiments.

  • Manufactured from ASTM Type 1 class A glass, ideal for variable temperature studies
  • Outer tube must be ordered separately depending on magnetic field strength

General applications include small volume NMR, external referencing, external locking and magnetic susceptibility determination.

Customers Who Bought This Also Bought