ContactUsLinkComponent
Mounting Rod, Material: Suprasil, Single Crystal, manufactured with the highest grade of synthetic quartz to eliminate E center defects and paramagnetic material contaminations, 2 sizes to choose from, Hexagonal rods are available upon request, Dimension: 4x5x1.5x2mm
# de catalogue 76460-272
Fournisseur:  SP Industries
Mounting Rod, Material: Suprasil, Single Crystal, manufactured with the highest grade of synthetic quartz to eliminate E center defects and paramagnetic material contaminations, 2 sizes to choose from, Hexagonal rods are available upon request, Dimension: 4x5x1.5x2mm
# de catalogue 76460-272
Fournisseur:  SP Industries
Customers Who Bought This Also Bought