ContactUsLinkComponent
VWR Semi Micro pH Elektrode Gel Glass Sm123
VWR® Universal pH Electrodes
# de catalogue 76460-474
Fournisseur:  VWR International
VWR Semi Micro pH Elektrode Gel Glass Sm123
VWR® Universal pH Electrodes
# de catalogue 76460-474
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Ø:
    12/6 mm
  • Description:
    Single junction, ceramic diaphram
  • Material:
    Glass
  • Cat. no.:
    76460-474
  • Connector:
    BNC, 1 m cable
  • Type:
    SM 123 (semi-micro)
  • Electrolyte:
    Gel
Spécifications
A propos de cet article
Laboratory and field electrodes for general applications. Supplied with a fixed cable or plug head, VWR® electrodes can be connected to most pH meters on the market.
  • Comfortable handling
  • Reliable, rapid readings
  • Gel or liquid electrolyte, glass or epoxy body shaft
  • Available with different connections: Plug head or fixed cable (1 metre) with either DIN or BNC type plug
All VWR electrodes are checked at two points during the manufacturing process and then lot stamped to ensure quality. They are supplied with a soaker storage bottle and cap to make sure the pH-sensitive glass and reference junction remain in top condition, and are instantly ready for use.
Delivery: Includes pH-electrode only.
Customers Who Bought This Also Bought