Spinner Turbine, Material: PEEK, Variable temperature: operation from -150 deg C to +200 deg C, Can be mixed with originals during sample changer operation, Weight compatible with room temperature spinners, Susceptibility matched to the originals, Dimension: 10 mm
# de catalogue 76460-018
Fournisseur: SP Industries
Customers Who Bought This Also Bought