Spinner Turbine, Material: PEEK, Cost effective compared to the full VT version for sample changers, Variable Temperature -150 to +200deg C with 5 mm PEEK exchangeable fingers, Can also be used for spinning experiments, Includes high temperature external O-rings at the top & bottom, Dimension: 3-5mm
# de catalogue 76460-014
Fournisseur: SP Industries
Customers Who Bought This Also Bought