ContactUsLinkComponent
NMR Sample Tube, Thin Wall Precision Screw-Cap with Short Valve, 500MHz, used in sample degasification these tubes can maintain vacuum quality of >10-4 torr. For better vacuum, please check our Pressure/Vacuum Tube, Tube comes complete with open top screw cap and septum, Size: 5 mmx7in long
SP Wilmad-LabGlass Screw-Cap NMR Tubes, SP Industries
# de catalogue 76459-862
Fournisseur:  SP Industries
SP Wilmad-LabGlass Screw-Cap NMR Tubes, SP Industries
# de catalogue 76459-862
Fournisseur:  SP Industries
Spécifications
  • Length:
    178 mm (7")
  • Description:
    Screw cap tube with short valve (17 mm)
Spécifications
A propos de cet article
These precision screw-cap NMR tubes are manufactured from the highest quality borosilicate glass and are ideal for sample degasification.
  • Maintains a vacuum quality of >10 to 4 torr
  • Ideal for sample degasification
  • Each screw-cap tube comes with one PTFE/silicone septum
PTFE/ rubber septums are inert to most solvents and many corrosive materials but not recommended for multiple punctures.
PTFE/silicone septums are inert to most organic solvents and compounds but not recommended for strongly corrosive materials. The septum remains reliable after multiple punctures.
Customers Who Bought This Also Bought