ContactUsLinkComponent
NMR Tube Cap, Material: Ethylene Vinyl Acetate, Colour: Sky Blue, are manufactured from molded Polyethylene depending on the tube OD, colors help track samples based on various reference solvents, avoid direct contact between the sample and disposable cap, long term storage over 2 hours, Size: 5mm
SP Wilmad-LabGlass Disposable NMR Tube Caps, SP Industries
# de catalogue 76459-678
Fournisseur:  SP Industries
SP Wilmad-LabGlass Disposable NMR Tube Caps, SP Industries
# de catalogue 76459-678
Fournisseur:  SP Industries
Spécifications
  • Color:
    Sky blue
  • Material:
    EVA (Ethylene vinyl acetate)
  • Cat. no.:
    76459-678
  • For Tube O.D.:
    5.0 mm
Spécifications
A propos de cet article
Manufactured from high quality polyethylene or ethylene vinyl acetate to ensure a tight seal.
  • Different colors help to track samples
  • Available in large quantities for high throughput laboratories
Caution: Please avoid using disposable NMR tube caps when CDCl₃ serves as the reference solution as the material(s) could be dissolved. For CDCl₃, we recommend PTFE tube caps.
Customers Who Bought This Also Bought