NMR Sample Tube, Heavy Wall Precision Pressure/Vacuum Valve, 450MHZ, for medium range pressure (<300 psi) and ultra vacuum (10-8 torr) experiments on the market, It is designed to connect to a 1/8in metal (SST or brass) vacuum line fittings or a rubber vacuum hose a glass connector, Size: 10mmx7in l
# de catalogue 76459-630
Fournisseur: SP Industries
Customers Who Bought This Also Bought