Back Ferrule, Material: Stainless Steel, For 1/8 OD Tubing
# de catalogue 76459-608
Fournisseur: SP Industries
Back Ferrule, Material: Stainless Steel, For 1/8 OD Tubing
# de catalogue 76459-608
Fournisseur: SP Industries
Customers Who Bought This Also Bought
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
220,41 $ to 875,50 $
Sodium azide ≥99.7%, OR, Macron Fine Chemicals™
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
350,13 $
Direct Thermal Cryotags, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
184,61 $ to 196,14 $
MinION™ Mk1C DNA/RNA Sequencing Device, Oxford Nanopore Technologies
Fournisseur: Oxford Nanopore Technologies
10 713,98 $