ContactUsLinkComponent
VWR* Examination Gloves, Material: Vinyl, Synthetic, Powder free, Non Sterile, Size: Medium
VWR® Vinyl Examination Gloves
# de catalogue 76459-216
Fournisseur:  VWR International
VWR* Examination Gloves, Material: Vinyl, Synthetic, Powder free, Non Sterile, Size: Medium
VWR® Vinyl Examination Gloves
# de catalogue 76459-216
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Glove Color:
    Natural
  • Glove size:
    M
  • Length:
    9.5"
  • Powdered/Powder-free:
    Powder-free
  • Disposable / reusable:
    Disposable
  • Latex/Latex-free:
    Latex-free
  • Sterility:
    Non sterile
  • Cat. no.:
    76459-216
Spécifications
A propos de cet article
This formulated blend of vinyl provides protection in low risk enviornments while maintaining protection, comfort, and dexterity.
  • Soft fit and feel
  • Latex-free and powder-free
  • DOP and DEHP (phthalate) free
  • Smooth inner coating for easy donning, and to help avoid irritation
  • Applicable for many industries
Suitable alternative for individuals with Type I allergies. Great for a wide range of applications including medical, examination, healthcare, nursing, laboratory, IT, general cleaning, and industrial use.
Meets ASTM D5250 standards for poly-vinyl chloride gloves for medical applications. Meets EN 374 and EN 455 standards. FDA 510k certified and meets food handling requirements.
Packaging: Pack of 100 gloves, case of 10 packs.
Customers Who Bought This Also Bought