Exam Gloves, 93-732, 1.5 AQL, Material: Nitrile, Color: Black, Chlorinated, Silicone Free, Textured Fingers, Beaded Cuff, Enhanced dexterity and tactility in a thin, yet strong design, Silicone free to ensure advanced product protection, Length: 9.6inch, Size: Large (8.5 - 9)
# de catalogue 76459-136
Fournisseur: Ansell Healthcare
Customers Who Bought This Also Bought