Autoclave Bottle, Phoenix, Fully compatible with 45 mm self-venting, rubber sealing cap, Autoclavable at 121deg C, Decades of proven and reliable use within gnotobiotic core facilities, High-purity, clear, USP/EP/JP, for use with 45 mm push-on rubber cap, Volume: 2000 ml
# de catalogue CA76459-164
Fournisseur: Kimble Chase
Customers Who Bought This Also Bought