Capillary Tool Kit, Make Life Easier (MLE), for TRACE 1300/1310 GCs, Includes: Capillary installation gauge for use with M4 ferrules, 1/16inch capillary installation tool, nylon tube brushes, stainless-steel wire tube brushes, open end wrench, Slide-lock tweezers, and Scoring wafers
# de catalogue 76457-574
Fournisseur: Restek
Customers Who Bought This Also Bought