Vial & Cap Kit, Includes 300ul, Fused Insert, Screw Top, Amber Autosampler Vials and 100 EA matching Screw Caps with White Silicone Rubber / Red PTFE Soft-Guard Septa, bonded in Blue Screw Caps
# de catalogue 76457-546
Fournisseur: MicroSolv Technology Corp
Customers Who Bought This Also Bought