ContactUsLinkComponent
VWR® Disposable Scalpels with Measurements
Catalogue # 76457-484
Fournisseur:  VWR International
undefined
VWR® Disposable Scalpels with Measurements
Catalogue # 76457-484
Fournisseur:  VWR International
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Disposable scalpel, sterile
  • Cat. no.:
    76457-484
  • Blade No.:
    #10
Spécifications
À propos de cet article
VWR® Sterile Disposable Scalpels are mounted on plastic handles with fluted finger-grips.
  • Wrapped in individual peel pouch
  • Sterilized by gamma radiation
  • Preassembled - Mounted on plastic handles with fluted finger-grips
Sterile. 10 per pack.
Certifications: ISO, GMP
Frequently Bought Together