ContactUsLinkComponent
VWR Scalpel, Knife Handle No 4
VWR® Scalpel Knife Handle
# de catalogue 76457-418
Fournisseur:  VWR International
VWR Scalpel, Knife Handle No 4
VWR® Scalpel Knife Handle
# de catalogue 76457-418
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • For:
    VWR 20 series
  • Length:
    12.07 cm (4.75")
  • Description:
    Scalpel knife handle, No.4
  • Cat. no.:
    76457-418
Spécifications
A propos de cet article
These high-quality scalpel knife handle fits the VWR® 10 and 20 series scalpel blades.
  • Secure blade fitment
  • Autoclavable/sterilizable
  • Ribbed/grooved
For classroom dissection, field-study, removal of small embedded objects and crafts.
Fits VWR carbon and stainless steel scalpel blades.
Certifications: ISO, GMP
Customers Who Bought This Also Bought