ContactUsLinkComponent
VWR Lab Coat SMS Blue No Pocket, Snap Front, Standard Collar, Open Cuffs X-Large Made in USA
VWR® SMS Lab Coats, Made in USA
Catalog # 76457-026
Supplier:  VWR International
VWR® SMS Lab Coats, Made in USA
Catalog # 76457-026
Supplier:  VWR International
Specifications
  • Clothing Size:
    XL
  • Garment application:
    Laboratory
  • Pockets:
    Without pockets
  • Color:
    Blue
  • Cat. no.:
    76457-026
Specifications
About this item
The VWR® Made in USA PPE line has been expanded to include SMS lab coats.
  • Multilayer SMS material protection
  • Light fluid and particulate barrier
  • Excellent breathability
  • Serged seams
  • Standard or knit collar options
  • Elastic, open, or knit cuff design
  • White or blue color options
Available in two colors and a variety of styles to meet virtually all disposable lab coat needs. Choose from different cuff options, such as knit, elastic, or no cuff. Additionally, different pocket options are available, including the popular no pocket choice.
Ordering information: Also available in the Made in USA PPE line are masks, coveralls, scrubs, and microporus lab coats.
Packaging: Packed 30 coats per case.
Frequently Bought Together