Polystyrene Storage Jars
# de catalogue 470330-298
Fournisseur: PLASTIC BOTTLES INC.
Spécifications
- Cap Size:89 mm
- Capacity:8 oz.
- Cat. no.:470330-298
Spécifications
A propos de cet article
Ideal for specimen storage.
- Clear polystyrene
- Wide-mouth
- Aligned with GLOBE protocols
Ordering information: Metal jar caps sold separately.
Customers Who Bought This Also Bought