Kit, Quick-Dna/Rna Magbead, Binding Capacity: 15 Ug Dna/Rna Per 30 Ul Magbinding Beads, Sample Sources: Any Cells, Solid Tissue, Whole Blood, Biological Fluids, Ffpe Tissue, Environmental (Plant/Seed), Swabs, Samples Stored In Dna/Rna Shield, Etc, Ngs-Ready, Versatile, Size: 96Preps
# de catalogue 76197-069
Fournisseur: Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.