Vwr* Balances, U-Series, With Table, Readability: 1UG, Touchless operation via IR proximity sensors, Internal and external calibration, Seven databases, provide unsurpassed performance ideal for any laboratory condition, offer a variety of standard features, such as GLP procedures, Capacity: 31gram
# de catalogue MP76446-826
Fournisseur: VWR International
Customers Who Bought This Also Bought