ContactUsLinkComponent
Vwr* Precision Balance, T-series, With Certifications, Voltage: 100 - 240 V, Plug Type: Nema 5-15R, Short Removable Draft Shield, Temperature Range: 10-40deg C, Readability: 0.0015 g, Repeatabilty: 0.001 g, Stabilization time: 2 s, Linearity: +/-0.003 g, platform size: 12.8x12.8 cm, capacity: 600 g
# de catalogue MP76300-072
Fournisseur:  VWR International
Vwr* Precision Balance, T-series, With Certifications, Voltage: 100 - 240 V, Plug Type: Nema 5-15R, Short Removable Draft Shield, Temperature Range: 10-40deg C, Readability: 0.0015 g, Repeatabilty: 0.001 g, Stabilization time: 2 s, Linearity: +/-0.003 g, platform size: 12.8x12.8 cm, capacity: 600 g
# de catalogue MP76300-072
Fournisseur:  VWR International
Customers Who Bought This Also Bought