Vwr* Precision Balance, B2-Series, with Draft Shield, With 200G weight, Temperature Range: 10-40deg C, Voltage: 100-240V, Readability: 1 mg, Repeatabilty: 1 mg, platform size: 128 x 128 mm, Linearity: 2 mg, External calibration, Forced compensation weighing technology, capacity: 400 g
# de catalogue MP75802-866
Fournisseur: VWR International
Customers Who Bought This Also Bought