ContactUsLinkComponent
Vwr* Carboy Assembly, Single Use, Material: Polypropylene, Color: Dark Amber, 83B Cap, TPE Tubing w/ Dip Tube, Gamma Sterilized, Autoclavable, USP Class VI, FDA Grade materials, Clear for high visibility of the product, High-impact strength, rectangular shape saves valuable bench space, Volume: 10L
# de catalogue 76449-458
Fournisseur:  VWR International
Vwr* Carboy Assembly, Single Use, Material: Polypropylene, Color: Dark Amber, 83B Cap, TPE Tubing w/ Dip Tube, Gamma Sterilized, Autoclavable, USP Class VI, FDA Grade materials, Clear for high visibility of the product, High-impact strength, rectangular shape saves valuable bench space, Volume: 10L
Vwr* Carboy Assembly, Single Use, Material: Polypropylene, Color: Dark Amber, 83B Cap, TPE Tubing w/ Dip Tube, Gamma Sterilized, Autoclavable, USP Class VI, FDA Grade materials, Clear for high visibility of the product, High-impact strength, rectangular shape saves valuable bench space, Volume: 10L
# de catalogue 76449-458
Fournisseur:  VWR International
Customers Who Bought This Also Bought