Vwr* Carboy Assembly, Single Use, Material: HDPE, 83B Cap, TPE Tubing w/ Dip Tube, Gamma Sterilized, Autoclavable, USP Class VI, FDA Grade materials, Clear for high visibility of the product, High-impact strength, Ergonomic shape, Leakproof, rectangular shape saves valuable bench space, Volume: 20L
# de catalogue 76449-418
Fournisseur: VWR International
Customers Who Bought This Also Bought