ContactUsLinkComponent
Freezer Box, Hinged Lid, Blue, 81-Place, Complete with Dividers, Fliptop, made of recyclable cardboard, with a liquid resistant outer coating, has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc.)
# de catalogue 76449-614
Fournisseur:  Chemglass
Freezer Box, Hinged Lid, Blue, 81-Place, Complete with Dividers, Fliptop, made of recyclable cardboard, with a liquid resistant outer coating, has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc.)
# de catalogue 76449-614
Fournisseur:  Chemglass
Customers Who Bought This Also Bought