Spinner Turbine, for Bruker Systems, Room Temperature Use, Color: White, Causes less probe insert damage by better insert sample control, Tachometer strip now with foil protection for improved life span, Can be mixed with originals during sample changer operation, Dimension: 10mm
# de catalogue 76449-898
Fournisseur: SP Industries
Customers Who Bought This Also Bought