ContactUsLinkComponent
Spinner Turbine, for Bruker Systems, Room Temperature Use, Color: White, Causes less probe insert damage by better insert sample control, Tachometer strip now with foil protection for improved life span, Can be mixed with originals during sample changer operation, Dimension: 3mm
# de catalogue 76449-884
Fournisseur:  SP Industries
Spinner Turbine, for Bruker Systems, Room Temperature Use, Color: White, Causes less probe insert damage by better insert sample control, Tachometer strip now with foil protection for improved life span, Can be mixed with originals during sample changer operation, Dimension: 3mm
# de catalogue 76449-884
Fournisseur:  SP Industries
Customers Who Bought This Also Bought