CD40 monoclonal Antibody, Clone: G28.5, Host: Mouse, Species reactivity: Human, Isotype: IgG1, Synonyms: TNFRSF5, Applications: Flow Cytometry, Functional Assays, reacts with human CD40, a 45-48 kDa glycoprotein that is a member of the TNF-receptor superfamily, size: 100ug
# de catalogue 76447-192
Fournisseur: Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.